Penetration Needle, Koehler
Catalog #: 52934-662
Supplier: Koehler
Small Business Enterprise
Specifications
- Description:Penetration Needle
- Cat. no.:52934-662
- Supplier No.:K20500-00000
Specifications
About this item
For testing of bituminous materials
Made from hardened stainless steel with a brass shank.
Certifications: Meets ASTM D5 specifications.
Customers Who Bought This Also Bought