AutoPak™ Power Units, Carver
Supplier: Carver
Certificates
About this item
A self-contained, electrically-operated automatic power unit available for retrofit to existing manual presses
It is equipped with a microprocessor control system and features a 1/4hp DC motor, two-stage hydraulic pump, externally adjustable pressure control, pressure gauge, and automatic cycle control. Two-hand “anti-tiedown” controls protect the operator. 115V.
Specifications
- Catalog No:
- 53880-070
- 53880-068
- 53880-066
- W×D×H:
- 50.8×58.4×61 cm (20×23×24")
- 50.8×58.4×61 cm (20×23×24")
- 50.8×58.4×61 cm (20×23×24")