ContactUsLinkComponent
Emergency Shower Tester and Eyewash Test Kit, Speakman®
Emergency Shower Tester and Eyewash Test Kit, Speakman®
Catalog #: VF-0950-SE
Supplier:  Speakman
Emergency Shower Tester
Emergency Shower Tester and Eyewash Test Kit, Speakman®
Catalog #: VF-0950-SE
Supplier:  Speakman
Specifications
  • Description:
    Shower and Eyewash test kit
  • Cat. no.:
    VF-0950-SE
  • Supplier No.:
    SE-950
Specifications
About this item
Consists of a vinyl-coated rayon sleeve/funnel supported by a handle to direct water flow, plus a 18.9 L (5 gal.) pail to catch water.
  • Hoop at end of sleeve fits all standard showers
  • Ideal for monthly inspections as required by good laboratory practices
Clear Plexiglas® test kit allows eyewashes to be tested for correct spray pattern.
Customers Who Bought This Also Bought