Disposable Pasteur Pipettes, Borosilicate Glass, Wheaton
Catalog #: 53499-630
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Specifications
- Cat. no.:53499-630
- Overall Length:14.6 cm (5¾")
- Supplier No.:357331
Specifications
About this item
For use in inoculating culture media and as a general sampling pipet.
Also used in applying samples in chromatography. Straight tip can be bent over a flame. Constriction near end to facilitate plugging. Pipettes have an Ext.Ø of 7 ±0.5 mm at the top.
Ordering information: Case of 1000 Pasteur Pipettes. Each case includes 4 inner cartons of 250 pipettes.
Customers Who Bought This Also Bought