AccuTec Pro® Hobby Blade #11
Catalogue # 77587-136
Fournisseur: Accutec Blades
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Coating:Oiled
- Material:Carbon steel
- Thickness:0.021"
- Environmentally Preferable:
- Cat. no.:77587-136
- Blade Edge:2 facet
Spécifications
À propos de cet article
AccuTec Pro® #11 Hobby blades are constructed from carbon steel for increased strength, durability, and sharpness. Precision acute point for fine cuts.
- Fits all standard #1 handles
- Excellent for precision cutting and trimming
- Made in USA
Packaging: Each box contains 100 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.