In-Transit Temperature Monitor/Recorder, Cold Chain Technologies
Catalog #: 33500-508
Supplier: Cold Chain Technologies
Small Business Enterprise
Specifications
- Temperature Range:–28°C to 38°C (–18°F to 100°F)
- Cat. no.:33500-508
- Supplier No.:TCHART-10-V
Specifications
About this item
Tamper-resistant, simple-to-operate, single-use, reliable, battery-powered chart recorder generates a permanent paper strip chart data record of temperature conditions and time duration for a period of 10 days or less
Each unit has a tamper-evident seal with a bar code and serial number. Requires no pre-conditioning of the indicator, product, or package, and provides visible start-up verification, Hazard Analysis Critical Control Points (HACCP) documentation, and NIST traceable calibration (±1°C).
Customers Who Bought This Also Bought