SP Bel-Art PiRack® Pipettor Holder System, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
CE Compliant
Certificates
About this item
This polypropylene system is strong enough to support the weight of even the heaviest pipettor.
- Polypropylene construction
- Strong and durable
- Utility tray included for holding small pipetting supplies
- Chemical-resistant
- Autoclavable
Hold a single pipettor with one clamp, or add a triple clamp or extenders to increase the number of holders at a work location. The utility tray, which mounts between two clamps, is ideal for storing pipetting supplies or other small items.
Attach to any shelf or benchtop edge up to 2.54 cm (1") thick using the large locking thumbscrew clamp. Supports many standard brands of pipettors. All components are chemical-resistant and autoclavable at 121°C (250°F).
Customers Who Bought This Also Bought