Needle Valves, WaterSaver Faucet
Supplier: WaterSaver Faucet
Certificates
About this item
Straight pattern laboratory needle valve is for gas/air/vacuum service.
Complete with 9.5 mm (3/8") IPS mounting shank, locknut, and washer. Maximum working pressure of 125 psi. Please specify service when ordering.
It has a removable serrated hose end and forged brass four-arm handle and valve body. Stainless steel seat and floating cone are removable/replaceable for infinite service life.
Caution: Not for use with water.
Customers Who Bought This Also Bought