To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Max. sample capacity:200 Samples
- Unit dimensions:27W x 31D x 7.5H cm (105/8 x 127/32 x 3")
- Voltage limit:170VDC
- Description:Ultra-Wide Mini Submarine Gel System
- Gel dimensions:23.5W x 15H cm (91/4 x 529/32")
- Cat. no.:82031-126
- Buffer Volume:850 ml
- Format:Mini
- Gel Dimensions:15 x 23.5 cm
- Supplier No.:JSB-96
Specifications
About this item
This electrophoresis system allows quick-screening of large quantities of samples with limited agarose usage
The system is ideal for PCR applications, RFLP analysis, and plasmid screening. Constructed of durable, three-piece cast acrylic. Microtiter plate format can run samples, standards, and blanks in under 90 minutes. Comb 82031-142 is compatible with most multichannel pipettors.