To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Unit dimensions:40W x 27L x 18H cm (153/4 x 105/8 x 71/8")
- Voltage limit:175VDC
- Description:HR2525 High-Throughput System
- Unit weight:2.2 kg (4.9 lbs.)
- Gel dimensions:25W x 25L cm (97/8 x 97/8")
- Cat. no.:14229-210
- Buffer Volume:1.5 L
- Gel Dimensions:25 x 25 cm
- Supplier No.:IB57000
Specifications
About this item
This large-format high throughput system is designed to run up to 200 samples on a single gel in less than 30 minutes
The vented lid reduces condensation, improving output resolution, and reduces heat build-up during electrophoresis, keeping the system running cool. The gravity feed electrode connector eliminates sample disturbance during setup and increases user safety by interrupting power when it is removed. The system is constructed of durable, heavy-duty, 1cm (3/8") thick acrylic. The multichannel pipettor format combs, available with 25 or 50 teeth, reduce the transfer time from plates to gel.
Ideal for running all types of precast gels.