MLA Digital Pipettors, Adjustable Volume, VistaLab
Supplier: VistaLab
Certificates
About this item
MLA digital pipettors are ideally suited for use in laboratories that perform tests requiring multiple or non-standard volumes.
- All models are engraved with serial numbers for improved traceability and calibration recordkeeping
- In-lab calibration is performed with minimal effort
- Manufactured from engineered aluminum alloy and stainless steel for maximum durability and performance
- Streamlined shape and smooth, single-stroke operation facilitate extended use
MLA digital pipettors are ideally suited for use in laboratories that perform tests requiring multiple or non-standard volumes.
Pipettors feature easy to read digital volume settings that are simply dialed in with click stops, providing fast and efficient volume selection. Low spring forces on all operating mechanisms reduce the fatigue and stress of repetitive procedures.
BodyTextAccessories: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.
Ordering information: More specifications are available in the Related Information section.
Customers Who Bought This Also Bought