Owl™ Sequencing Gel Casters, Model SGC, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Cast bubble-free sequencing gels in under three minutes.
- Capillary action draws gel between the plates
- Surface tension eliminates leaks
- No tape or special assembly is needed
Easy-to-use gel casters are adjustable from 20 cm (77/8") wide to 42 cm (161/2") wide.
Ordering information: Systems include adjustable casting platform, width adjustment knobs, and 12 tempered steel binder clamps.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.