Axygen® Automation Tips for Tecan®Corning
Supplier: Corning
Certificates
About this item
PP, with or without filter, sterile or non sterile.
- Black tips are conductive for liquid level-sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Specifications
- Catalog No:
- 89125-544
- 89087-538
- 89080-490
- 89040-090
- 89040-092
- 89040-086
- 89040-082
- 89040-084
- 89040-074
- 89040-060
- CA10030-828
- CA10030-824
- CA10030-826
- 89087-340
- CA10030-822
- 89080-504
- 89080-502
- 89080-500
- 89080-498
- 89080-496
- 89080-494
- 89040-072
- 89040-068
- 89040-070
- 89080-506
- 89040-062
- 89040-066
- 89040-104
- 89040-058
- 89040-100
- 89040-102
- 89040-096
- 89040-098
- 89040-064
- 89040-094
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Related Information
Certificates
Search by Lot Number89040-058_18319022 89040-060_05919008 89040-062_06619005 89040-062_06919011 89040-062_07319010
Conformity declaration
More
Axygen® 1000 µl Automation Tips for Tecan® Freedom EVO® 200 with LiHa head – Precision and Accuracy Axygen® 200 µL Automation Tips for Tecan® Freedom EVO® 200 with LiHa head – Precision and Accuracy Axygen® 50 µl Automation Tips for Tecan® Freedom EVO® 200 with LiHa head – Precision and Accuracy Axygen® Automation Tips Performance Testing for Axygen® Automation Tip (EV-100-R)
More
Customers Who Bought This Also Bought