ContactUsLinkComponent
MASTERBLOCK® 1536-Well Deep Well Plates, Polypropylene, Greiner Bio-One
MASTERBLOCK® 1536-Well Deep Well Plates, Polypropylene, Greiner Bio-One
Catalog #: 82051-410
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
MASTERBLOCK 1536-Well Deep Well
MASTERBLOCK® 1536-Well Deep Well Plates, Polypropylene, Greiner Bio-One
Catalog #: 82051-410
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Specifications
  • Color:
    Clear
  • Working Well Volume:
    3 - 15 µl
  • Maximum Well Volume:
    18 µL
  • Supplier No.:
    782270
Specifications
About this item
These conical-bottom microplates feature square-round internal well geometry to minimize wicking and air bubble formation.
  • Free of RNase, DNase, and DNA-polymerase inhibitors
  • Polypropylene construction
  • Low-binding and biologically inert
  • Autoclavable
Deep well microplates are manufactured of ClearChoice™ virgin resin in molds specifically engineered for polypropylene
Microplates are RNase-free, DNase-free, nonpyrogenic, and free of DNA-polymerase-inhibitors.
Produced with uniform dimensions and tolerances to meet SBS recommendations for compatiblity with equipment and robotics. Polypropylene is biologically inert, low binding, autoclavable, and resistant to many standard lab chemicals.
Customers Who Bought This Also Bought