About this item
Designed for experiment methods optimization and large volume electroporation
The system also features integrated electrodes that eliminate cross-contamination. Plates allow users to prepare and run an entire set of experiments at once. This approach minimizes sample and waste materials. Multi-Well plates can be electroporated in seconds.
Plate handlers are designed to operate in space-constrained areas such as laminar flow hoods. They feature gold-plated contacts to mate with the electroporation plate and connectors for standard pulse generators. Economical plates require no expensive reagents, provide reliable results, and ensure that experiments progress more quickly. The 25-well plates feature a 5x5 format, while the 96-well plates feature a standard 8x12 format.
Both the HT-100 Advanced Electroporation Plate Handler and the HT-200 Advanced Electroporation Plate Handler offer manual pulse switching technology, and are designed to work with 96-well plates. They are supplied with an adapter to accommodate 25-well plates as well with the ECM® 630 generator (compatible with ECM® 830).
Applications include the transfection of mammalian cells, the transformation of bacteria and yeast, and cDNA library and drug screening. High Throughput Systems offer multi-well electroporation technology for processing multiple samples in seconds. By utilizing HT Multi-Well plates instead of traditional cuvettes, users can transition from a single cuvette to either a 96-well or 25-well electroporation plate through the use of one simple plate handler. Plate handlers enable quick and efficient optimization of electrical and biological parameters associated with any electroporation experiment. Once optimized, samples may be rapidly processed, increasing yields and saving valuable time and money.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 47746-064
- 47746-075
- 47746-062
- W×D×H:
- 31.75×31.1×14 cm (121/2×121/4×51/2")
- 31.75×31.1×14 cm (121/2×121/4×51/2")
- Pulse interval:
- 100 msec to 10 sec
- 100 msec to 10 sec
- Capacitance in high voltage mode:
- 110 µF
- 110 µF
- Ambient operating temperature:
- 5...40 °C (41...104 °F)
- 5...40 °C (41...104 °F)
- Amperage:
- 500 A limit low voltage mode, 100 A limit high voltage mode
- 500 A limit low voltage mode, 100 A limit high voltage mode
- Input:
- 110V/220V Universal
- 110V/220V Universal
- Rapid charging:
- <13 seconds
- <13 seconds