Order Entry
ContactUsLinkComponent
Pierce™ Modified Lowry Protein Assay Kit, Thermo Scientific
Pierce™ Modified Lowry Protein Assay Kit, Thermo Scientific
Catalog #: CAPI23240
Supplier:  Thermo Scientific
CAS Number:  
Modified Lowry Protein Assay Kit
Pierce™ Modified Lowry Protein Assay Kit, Thermo Scientific
Catalog #: CAPI23240
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Modified Lowry Protein Assay Kit
  • Cat. no.:
    CAPI23240
  • No. of Assays/Kit:
    480 standard tube assays or 2,400 microplate assays
  • Supplier No.:
    23240

Specifications

About this item

The Modified Lowry Protein Assay Kit is the preferred colorimetric assay for quantifying total protein concentration.

  • Ready-to-use reagent for the loyal Lowry method user
  • Preformulated cupric sulfate-tartrate reagent stable for one year at room temperature
  • Linear results from 1 to 1,500 µg/mL for BSA
  • Adaptable to microplates
  • Less protein:protein variation than dye-binding methods

The sensitivity of the Modified Lowry Protein Assay is greatly enhanced over that of the biuret reagent. The working range of the method extends from 5 to 2,000 mg/mL. Using cold Modified Lowry Protein Assay Reagent will result in low absorbance values.

Delivery: Kit includes Modified Lowry Protein Assay Reagent (480 mL), 2 N Folin-Ciocalteu Phenol Reagent (50 mL), Albumin Standard Ampoules, 2 mg/mL, (10×1 mL).
Customers Who Bought This Also Bought