About this item
Sealing films are ideal for 96-well, 384-well, deep well, and other assay and PCR plates.
Aluminum Seal films maintain an excellent seal on microplates for a wide temperature range. Ideal for light-sensitive samples. Its uniformly applied adhesive provides optimal well sealing. When used in conjunction with Axygen compression mat, seals during thermocycling process. Color: reflective. Temperature range: –80 to 97°C (–112 to 207°F).
AxySeal films are polyester-based films with an acrylic adhesive ideal for ELISA and enzyme immunoassay applications. Color: clear. Temperature range: –40 to 65°C (–40 to 149°F).
UltraClear film is an exceptionally clear film specifically designed for meeting the optical requirements of Real-Time PCR. With a unique pressure-sensitive adhesive, this film has no tacky residue that can interfere with optical analysis. Color: clear. Temperature range: –40 to 65°C (–40 to 149°F).
Breathable films allow effective gas exchange for cellular/bacterial cultivation while preventing contamination. Color: clear. Temperature range: room temperature.
Specifications
- Catalog No:
- 47749-926
- 47749-924
- 47743-943
- 89080-530
- 47734-817
- 89080-528
- 10011-117
- 10011-119
- 10011-121
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
Specifications
Related Information
Certificates
Search by Lot Number10011-117_35119961 10011-121_02621795 89080-528_02321781 89080-530_32819025
Conformity declaration
Corning Axygen Automation-Compatible PCR Microplate Flyer Corning Axygen® 96-well Deep Well Microplates and Sealing Solutions Corning Axygen® Brand Equipment Product Selection Guide Corning Axygen® MaxyGene II Thermal Cycler Corning ELISA Workflow Guide
Product Literature
Corning New Lab Startup Program Corning Special Offer