Microtiter® 96-Well UV Microplates, Thermo Scientific
Catalog #: 14227-680
Supplier: Thermo Fisher Scientific
Specifications
- Description:Microtiter® 96-Well UV Microplates
- Cat. no.:14227-680
- Supplier No.:8404
Specifications
About this item
These flat-bottom UV plates offer a low background and higher throughput at wavelengths of 260 and 280nm
Ideal for nucleic acid and protein determinations. Plates are robotic-friendly with a standard 96-well footprint and are perfect for use with any standard UV/Vis spectrophotometer.
Customers Who Bought This Also Bought