About this item
Finnpipette™ F2 multichannel pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures.
Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision. Each lightweight device offers remarkably smooth pipetting action. Pipettors are fully autoclavable and easy to service. An enlarged digit display improves ease of use, and color coding simplifies instrument selection.
- Fully autoclavable
- An enlarged digit display improves ease of use
- Color coding simplifies instrument selection
- Easy to service
Finnpipette™ F2 multichannel pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures.
Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision. Each lightweight device offers remarkably smooth pipetting action.
These pipettors combine established Finnpipette™ features with state-of-the-art innovations to create a reliable and ergonomic high-performance device.