Order Entry
Canada
ContactUsLinkComponent
Microlite™ Microtiter® 96-Well Plates and Strips, Thermo Scientific
Microlite™ Microtiter® 96-Well Plates and Strips, Thermo Scientific
  62402-974
undefined
Microlite™ Microtiter® 96-Well Plates and Strips, Thermo Scientific
  62402-974
 :  7417-12

 

  • Format:
    Plate
  • No. of wells:
    96
  • Well Volume:
    0.33 mL
  • Description:
    Microlite™ 2
  • Size:
    12.8 x 8.6 cm (5 x 33/8")
  • Cat. no.:
    62402-974
  • Supplier No.:
    7417-12

 

 

These flat-bottom plates were developed for luminescence applications, including PCR quantification, report gene detection, phagocytosis and luminescent immunoassay.

Made of a densely pigmented, white polystyrene formulation developed for luminescent assays, Microlite™ 1 is ideal for applications requiring high reflectivity and minimal crosstalk. Microlite™ 2, irradiated, is ideal for applications requiring higher binding with the same high reflectivity and minimal crosstalk.