ThinCert™ Multiwell Plates, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Optimized for use with ThinCert™ cell culture inserts, these 6- and 12-well plates feature deep wells that allow for an increased volume of medium in air-lift culture.
- Stackable
- Sterile
- Flat-bottom wells also feature alphanumeric labeling
- Supplied with lids
This results in fewer medium changes and a reduced amount of manual work. Additionally, this ensures that the medium conditioning lasts longer and an improved tissue quality is acheived. Stackable plates are equipped with notches that secure ThinCert™ cell culture inserts into a stable position throughout cell culture processes. Flat-bottom wells feature alpha-numeric labeling. Supplied with lids.
Ordering information: For additional information on ThinCert™ cell culture inserts, see the 82050-010 series.
Related Information
Certificates
Search by Lot NumberAM-657110.pdf
Technical Drawing
AT-cokultur application note.pdf
Additional Product Details
Customers Who Bought This Also Bought