Falcon® HTS 24-Multiwell Insert Systems, Corning
Supplier: Corning
Certificates
About this item
The Falcon® HTS 24-multiwell insert system is designed to automate many commonly used membrane-based cell assays and increase the productivity and throughput of these assays in the drug discovery process. Available in a choice of membrane pore sizes (1, 3 and 8 µm), 24-multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability and absorption studies (Caco-2 cells), chemotaxis, cell migration and invasion assays.
- All the benefits of individual cell culture inserts in an automation-friendly format that is compatible with most robotic and fluid handling equipment
- Handle 24 inserts simultaneously, as all 24 inserts are part of a single unit that is compatible with Falcon® 24-well plates and feeder tray
Pore density: 1 µm insert (pores/cm²): 1.6×10⁶
Pore density: 3 µm insert (pores/cm²): 8.0×10⁵
Pore density: 8 µm insert (pores/cm²): 1.0×10⁵
Customers Who Bought This Also Bought