About this item
Ultralab™ systems, composed of Ultrareservoir™ containers and Ultrapump™ II pumps, are designed as a rapid, efficient replacement of dialysis for buffer exchange and desalting of macromolecules.
- Efficient replacement of dialysis
- Airtight lid
- Siphoning action
Systems are specifically designed for use with Ultrasette™ lab tangential flow devices (see 29300-988 series).
Ultrapump™ II has a 0 to 600 rpm rated, ¹/₁₀ hp motor with continuous-duty rated drive, and 0 to 10 dial speed control. Pump size: 16.3×34×13.7 cm (6¹³/₃₂×13¹³/₃₂×5¹³/₃₂"). Maximum pressure: 2.8 bar (40 psi) intermittent, 1.8 bar (25 psi) continuous. Min. recirculation volume: 20–30 ml. Ultrareservoir™ is constructed from acrylic with EPR lid O-ring, stainless steel gauge, and polypropylene valves, barb connectors, and lid.
Airtight lid and siphoning action maintain constant sample volume and eliminate the need for multiple buffer exchanges. Sample port allows maximum sample recovery without decanting. Peristaltic pump action excludes air in the system, reduces foaming, and minimizes shear of sensitive molecules.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA29301-084
- CA29301-082
- Type:
- 5 L
- 2 L
- Dimensions:
- 20.3 x 20.3 x 27.9 cm (8 x 8 x 11")
- 15.2 x 15.2 x 22.9 cm (6 x 6 x 9")
- Hold-up volume:
- \<5mL
- \<2 mL
- Inlet/outlet connection:
- 6.4 mm (1/4")
- 6.4 mm (1/4")