To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:4.5 mm (³/₁₆"), Tip
- Length:150 mm (overall), 90 mm (tip)
- Max. I.D.:0...60 °C
- Temperature range:0...60 °C
- Cat. no.:14011-002
- pH Range:0 - 14
- Body Material:Glass
- Internal Reference:Ag/AgCl
- Junction:Ceramic
- Connector:BNC
- Supplier No.:9110DJWP
Specifications
About this item
This double junction pH electrode with semi-micro tip measures samples as small as 0.2 ml.
- With waterproof BNC connector
- Ag/AgCl double junction
- Refillable
Refillable semi-micro glass pH electrode with a waterproof BNC connector and 1 m (39 ³/₈") cable. For use in TRIS buffers and with proteins. Works with any pH meter with a BNC connector. Mercury-free and environmentally safe.