Hazardous Materials Shippers, Safety-Coated Glass, UN Marked, Qorpak®
Supplier: Qorpak
Certificates
About this item
Individual convenience packs suitable for shipping hazardous materials.
Certified packaging is supplied complete with safety-coated glass container, Thermoset closure with foam-backed fluoropolymer resin liner, corrugated outer packaging, cushioning material, absorbent, and tape.
- Plastic coated narrow-mouth bottles for liquids
- Plastic coated wide-mouth bottles for solid and viscous materials
- 120mL shippers are not plastic coated
Certified packaging is supplied complete with safety-coated glass container, Thermoset closure with foam-backed fluoropolymer resin liner, corrugated outer packaging, cushioning material, absorbent, and tape.
Meet UN 4GV requirements (labels required).
Warning: Shipper must not make any substitutions. Failure to heed this warning would invalidate package compliance.
Customers Who Bought This Also Bought