Falcon® Petri Dishes, Tight-Fit Lid, Sterile
Supplier: Corning
Certificates
About this item
Dishes are constructed with crystal-grade polystyrene and a flat bottom surface for distortion-free optics
- Flat, distortion free optics
- Stacking rings allow for easier stacking and handling
- Durable construction for stable dish manipulation
Small diameter saves media and space. Easily sealed with tape for collecting and transporting specimens. Polystyrene. Sterilized by gamma irradiation. Packed 20 per bag.
Related Information
Certificates
Search by Lot Number25373-041_9260010 25373-085_0001014 25373-085_9078012 25373-085_9246004 25373-085_9249002
Conformity declaration
More
Customers Who Bought This Also Bought