Nunclon™ Delta MultiDishes, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These polystyrene dishes with flat-bottom wells incorporate the Nunclon™Δ treated surface for optimal cell attachment and growth
Lids are keyed for a one-way fit. Rings in the lids reduce evaporation and condensation. Writing areas on both lids and dishes and individual alphanumeric markings allow easy identification of samples. Nontoxic and certified nonpyrogenic. Stackable.
Related Information
Certificates
Search by Lot Number62407-068_1252991 62407-068_1252992 62407-332_162259 62407-338_162066 62407-338_162367
Conformity declaration
More
Thermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Customers Who Bought This Also Bought