VWR® Drying Tubes
Supplier: VWR International
Certificates
About this item
Ideal for drying gases, these polyethylene drying tubes have two push-on ends for easily filling with drying agents.
- Polyethylene construction
- Tapered
- Accept tubes with 8, 9, and 10 mm diameters
Crest ext.Ø.: 7.8 / 10 mm (⁵/₆ / ⅜").
Customers Who Bought This Also Bought