Axygen® MAXYMum Recovery™ PCR Tubes, Corning
Supplier: Avantor
Certificates
About this item
Designed for applications requiring high accuracy and reproducibility.
- Domed and flat tops are pierceable to reduce the risk of contamination
- Certified to be safe from RNase, DNase, and pyrogens
- Autoclavable under normal autoclaving guidelines
Thumb-friendly caps have large seals and full-length flanges, increasing pressure tolerance and minimizing sample evaporation during thermal cycling.
Thin, uniform wall thickness reduces insulating effect of tube. Constructed using diamond-polished molds to eliminate the surface occlusions and cavities that cause sample retention. Unique polymer prevents DNA, RNA, and protein adherence. Manufactured without secondary substances such as silicone, which can leach into samples.
Specifications
- Catalog No:
- 22234-054
- 22234-056
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa