SP Wilmad-LabGlass Solvent Jet Washers for NMR Tubes, Economy, SP Industries
Supplier: SP Industries
Certificates
About this item
Reliable for over 40 years, this simple design washing unit is the most economical for single tube cleaning.
- Each unit only accommodates NMR tube with Ext.Ø and length as indicated in specifications
- Requires filter flask (sold separately; 500 ml recommended)
Customers Who Bought This Also Bought