About this item
Measure sodium in aqueous solutions quickly, accurately, and economically with Orion™ Sodium Electrodes
A unique redox internal system with an Orion™ ROSS™ electrode provides faster response, greater stability, and better accuracy than conventional sodium electrodes.