Orion™ Iodide Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These electrodes are designed for measuring iodide ions in quickly, simply, accurately, and economically.
- Stable, drift-free potentials
- Sure-Flow® junction allows easy cleaning and is easy to use in tough samples
- When conventional electrode junctions clog, the Sure-Flow® junction can be opened and flushed clean simply by pressing the cap
- Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow® junction design
- Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes
Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.