About this item
Quickly, simply, accurately, and economically measure free fluoride ions in aqueous solutions with the combination Orion™ Fluoride Electrodes.
Measure free fluoride ions in aqueous solutions with the Orion™ Fluoride Electrodes. The Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples. The free flowing Sure-Flow™ junction assures stable, drift-free potentials. The half cell electrode requires a separate reference electrode. The reference electrode 900100 is recommended for use with thalf cell electrodes 9409BN and 9409SC.
Temperature range:
Continuous: 0 to 80 °C
Intermittent: 0 to 100 °C