VWR® Culture Tubes, Colored PP
Supplier: VWR International
Certificates
About this item
Disposable, non sterile, endotoxin-free tubes are made of ultra-clear resin.
- Ultra-clear resin fabrication
- Autoclavable
Disposable, non sterile, endotoxin-free tubes are made of ultra-clear resin.
Colored for easy identification of samples.
Specifications
- Catalog No:
- 60818-433
- 60818-434
- 60818-436
- 60818-438
- 60818-432
- 60818-437
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
Specifications
Related Information
Certificates
Search by Lot Number60818-436_942C2-942 60818-437_942C2-942 60818-438_950C2-950
Conformity declaration
Customers Who Bought This Also Bought