Orion™ Calcium Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Measure calcium in aqueous solutions simply, accurately, and economically with Orion™ Calcium Electrodes.
- Free-flowing Sure-Flow junction assures stable, drift-free potentials
- A separate reference electrode is unnecessary, making this electrode convenient to use with small samples
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.