VWR® Round-Bottom Tubes, PTFE
Supplier: VWR International
CE Compliant
Certificates
About this item
Round-bottom tubes are designed with good temperature and chemical resistance.
- Durable pure PTFE construction
- Sturdy, heavy wall construction
- Inert and autoclavable to 280 °C (536 °F)
Isostatically molded pure, FDA approved PTFE.
Customers Who Bought This Also Bought