VWR® Peg Racks
Catalog #: 82024-496
Supplier: VWR International
Specifications
- Color:White
- No. of Holes:96
- Cat. no.:82024-496
- For Tube O.D.:10 - 13 mm
Specifications
About this item
Ideal for bench work or storage, these polypropylene racks are reinforced with fiberglass to make them strong and versatile.
- Polypropylene construction
- Fiberglass reinforcement
- Autoclavable
Racks feature embossed indexing, ribbed grips and drainage holes. Spacious peg design allows easy viewing of tube contents. Tubes may be placed in the rack upside down for drying. Pegs remain stiff and straight through repeated autoclaving.
Customers Who Bought This Also Bought