ContactUsLinkComponent
VWR® Magnetized Pipette Rack
VWR® Magnetized Pipette Rack
Catalog #: 82007-220
Supplier:  VWR International
VWR* Magnetized Pipet Rack
VWR® Magnetized Pipette Rack
Catalog #: 82007-220
Supplier:  VWR International
Specifications
  • Dimensions:
    29W x 8.5D x 41H cm (111/2 x 33/8 x 16")
Specifications
About this item
Durable blue ABS plastic storage rack features circular magnets for secure mounting to metal fume hoods, refrigerators, file cabinets, and more.
  • Durable construction
  • Blue color
  • Magnetized
Four angled compartments accommodate 0.1 mL to 25 mL pipettes. Includes two movable attachments to hold pipetting devices. Attachment dimensions: 10.5Lx3Hcm (41/8x11/4").
Customers Who Bought This Also Bought