Whatman™ Cellulose Soxhlet Extraction Thimble, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems
- Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
- Suitable for temperatures up to 120°C (248°F)
- Excellent mechanical strength and retention
- Wall thickness of standard thimbles is 1 to 1.5 mm to support application need
- Option for double thickness for added rigidity
Standard extraction thimbles have a wall thickness of 1 to 1.5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.
Used in air and waste gas analysis.
Related Information
Certificates
Search by Lot Number27730-142_1694396902 27732-065_1713652101 CA27730-161_1695029401
Conformity declaration
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Environmental testing for water air and soil Brochure (US)
Product Literature
Customers Who Bought This Also Bought