Specifications
- Description:Cytoseal™ XYL Mounting Medium
- Size:118.3 mL (4 oz.)
- Cat. no.:CA48212-196
- Supplier No.:8312-4
Specifications
About this item
Inhibit the fading or yellowing of stained specimens with specially formulated Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ XYL mounting media.
- Xylene-based rapid drying allows microscopic examination of slides soon after application
- Simple dispensing
Inhibit the fading or yellowing of stained specimens with specially formulated Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ XYL mounting media.
This xylene-based mounting medium is made of high-quality synthetic resins and contains an antioxidant to inhibit stain fading and yellowing of specimen. Rapid drying medium allows specimen examination soon after application. Bottle features a twist-open cap for simplified dispensing.