To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:2.5 ml
- Enzyme name:Coagulase
- Cat. no.:CA90003-150
- Supplier No.:240826
Specifications
About this item
Lyophilized rabbit plasma
Contains 0.15% EDTA and 0.85% sodium chloride. Coagulase binds plasma fibrinogen, causing staphylocci associated with acute infection to agglutinate. This enzyme is recommended for the direct tube test. It reconstitutes to 15mL.