Nunc® Lab-Tek™ Chamber Slide™ Systems, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Systems feature a removable polystyrene medium chamber so cells can be grown, fixed, and stained on the same slide.
- Removable media wells are attached to either a soda-lime glass microscope slide or a solvent-resistant Permanox® plastic slide by a medical-grade silicone gasket
- A convenient writing surface is easily marked, providing positive sample identification
- Multiwell design allows researchers to analyze different specimens, immunochemicals, or stains on one slide without transfer from other vessels
Tested for quality and sterility.
Packaging: Packed sterile, 8 slides per incubator tray.
Caution: Do not use mounting media containing toluene or xylene on Permanox slides.
Related Information
Certificates
Search by Lot Number62407-305_1246627
Conformity declaration
Thermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Customers Who Bought This Also Bought