Mod-Rack™ Pegboard, Interdyne
Supplier: Interdyne
Certificates
About this item
Stainless steel pegboard features interchangeable white polypropylene pegs, integral drip trough, and clear plastic drain tube
Completely autoclavable and stain-resistant. Wall bracket included.
Ordering information: Accessories including a funnel rack, drain basket, screen insert, and pipet rack are also available; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought