VWR® Premium VistaVision™ Microscope Slides, Color-Coded
Supplier: VWR International
CE Compliant
Certificates
About this item
Bright printed colors of microscope slides provide an ideal marking area and enable easy identification.
- Manufactured in Germany using high-quality, white soda lime glass
- Pre-cleaned and ready for use
- Autoclavable
- Color-coded ends facilitate sorting and identification
- Uniform size and thickness
Ultra-thin color frosted end facilitates processing on automated systems.
Customers Who Bought This Also Bought