Corning® Plain Micro Slides, Corning
Supplier: Corning
Certificates
About this item
For use in nonsterile cytology and pathology applications
Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging.
Ordering information: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.
Customers Who Bought This Also Bought