Super Erecta Shelf® Lab Animal Research (LAR) Carts, Metro™
Supplier: Metropolitan Wire
Certificates
About this item
One and two-tote capacity carts for storing and transporting animal feed
- Durable, stackable white totes provide clean storage for animal feed
- Snap-on covers keep tote contents secure
- Wire cover holder provides a convenient storage option for tote box covers when not in use
- Cover holder attaches easily to wire shelf and takes up minimal space
- Stainless steel finish provides superior corrosion resistance
Compact footprint allows carts to maneuver in and around tight lab spaces. Super Erecta Shelf construction offers ease of assembly, absolute rigidity, and quick and simple relocation of wire shelves. Open construction minimizes dust accumulation and maximizes air circulation.
Dimensions - tote bottom: 21 x 16"
Dimensions - tote top: 221/2" x 171/2"
Tote height: 12"
Ordering information: Cart 82000-964 comes with one tote, one snap-on cover, one cover holder, and four light-duty, 10.2cm (4") swivel casters with resilient rubber tread. Cart 82000-966 comes with two totes, two snap-on covers, one cover holder, and four heavy-duty, 12.7cm (5") swivel casters with polyurethane tread. All casters include donut bumpers.
Customers Who Bought This Also Bought