PIG® Absorbent Pillow, New Pig
Supplier: 2 D 2 S.A. DE C.V. MX
Certificates
About this item
Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids. Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated. Cellulose filler is highly absorbent for long-term use or big spill cleanup. Compact design is great for catching persistent drips and leaks in small spaces or adding absorbency for spill response.
- Catches up to 7.6L (2gal.) of oils, coolants, solvents, and water
- Fast-wicking cellulose filler absorbs liquids instantly
- Tough polypropylene gray cover resists tears, ruptures, and rough handling
- Fits into small spaces
- Can be incinerated after use to reduce waste
Absorbs and retains oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Recycled content is Excellent when green products are desired or required. Can be incinerated after use to reduce waste or for fuels blending.
Customers Who Bought This Also Bought