Gold Seal® Cover Glasses, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Protect and preserve specimens on slides with Gold Seal® Cover Glasses, made of noncorrosive borosilicate glass.
- Noncorrosive, nonfogging glass
- Provides exceptional quality and superior moisture-resistance barrier packaging
- Sealed in a moisture-resistant plastic case that converts to a 1-piece slide file with clear cover
Ordering information: Packaged in a plastic, 100-piece slide file.
Customers Who Bought This Also Bought