Nunc® Lab-Tek™ SlideFlask, Sterile, Thermo Scientific
Catalog #: CA62407-355
Supplier: Thermo Fisher Scientific
Specifications
- Well Area:9.0 cm2
- Description:SlideFlask
- Size:18W x 50L mm
- Working Volume:2.5 - 5.0 ml
- Supplier No.:170920
Specifications
About this item
This SlideFlask is a sealable yet accessible pressure-resistant culture vessel/microscope slide
Note: For processing, the manufacturer recommends using methanol based solutions and aqueous coverslip reagents.
Polystyrene slide is tissue culture treated for anchorage dependent cells. Sonically welded polystyrene components provide leak resistance without gaskets or adhesives. Units snap apart easily to allow access to the slide for analysis.
Ordering information: Supplied sterile, complete with cap. Coverglass must be ordered separately.
Customers Who Bought This Also Bought