Nunclon™ Delta Culture Slides, Sterile, Thermo Scientific
Catalog #: CA62407-360
Supplier: Thermo Fisher Scientific
Specifications
- Material:Permanox Plastic
- Cat. no.:CA62407-360
- Supplier No.:160005
Specifications
About this item
These 25x75mm slides are Nunclon™Δ treated for tissue culture growth and attachment
These 25x75mm slides are Nunclon™Δ treated for tissue culture growth and attachment
Choose from Permanox® or polystyrene. Permanox® slides are manufactured from a chemically resistant plastic that does not interfere with fluorescent labels.
Ordering information: Packaged 20 per bag.
Customers Who Bought This Also Bought